missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-acetyl-L-lysine 98.0+%, TCI America™

Catalog No. p-7130042
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10411G 1 g
F10415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10411G Supplier TCI America Supplier No. F10411G

Chemical Identifiers

CAS 159766-56-0
Molecular Formula C23H26N2O5
Molecular Weight (g/mol) 410.47
MDL Number MFCD00077409
InChI Key HQLBYVWJOXITAM-RNZRUAGMNA-N
Synonym fmoc-lys ac-oh, fmoc-n'-acetyl-l-lysine, nalpha-fmoc-nepsilon-acetyl-l-lysine, l-lysine, n6-acetyl-n2-9h-fluoren-9-ylmethoxy carbonyl, 2s-6-acetamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoc-lys ac, fmoc-lysine ac-oh, fmoc-l-lys ac-oh, ksc005q1n, fmoc-n-epsilon-acetyl-l-lysine
PubChem CID 7018846
IUPAC Name (2S)-6-acetamido-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
SMILES CC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 164°C
Color White
Quantity 1 g
Formula Weight 410.47
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-acetyl-L-lysine
TSCA No
Recommended Storage Freezer