missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-citrulline 98.0+%, TCI America™
$42.87 - $128.62
Chemical Identifiers
| CAS | 133174-15-9 |
|---|---|
| Molecular Formula | C21H23N3O5 |
| Molecular Weight (g/mol) | 397.431 |
| MDL Number | MFCD00151943 |
| InChI Key | NBMSMZSRTIOFOK-SFHVURJKSA-N |
| Synonym | fmoc-cit-oh, fmoc-l-citrulline, nalpha-fmoc-l-citrulline, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-ureidopentanoic acid, l-ornithine, n5-aminocarbonyl-n2-9h-fluoren-9-ylmethoxy carbonyl, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-citrulline, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-ureido-pentanoic acid, 2s-5-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-citoh, pubchem19002 |
| PubChem CID | 2756127 |
| IUPAC Name | (2S)-5-(carbamoylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid |
| SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCNC(=O)N)C(=O)O |
Chemical Identifiers
| 133174-15-9 | |
| 397.431 | |
| NBMSMZSRTIOFOK-SFHVURJKSA-N | |
| 2756127 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCNC(=O)N)C(=O)O |
| C21H23N3O5 | |
| MFCD00151943 | |
| fmoc-cit-oh, fmoc-l-citrulline, nalpha-fmoc-l-citrulline, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-ureidopentanoic acid, l-ornithine, n5-aminocarbonyl-n2-9h-fluoren-9-ylmethoxy carbonyl, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-citrulline, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-ureido-pentanoic acid, 2s-5-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-citoh, pubchem19002 | |
| (2S)-5-(carbamoylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid |
Specifications
| 133174-15-9 | |
| C21H23N3O5 | |
| 1 g | |
| NBMSMZSRTIOFOK-SFHVURJKSA-N | |
| (2S)-5-(carbamoylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid | |
| 2756127 | |
| ≥98.0% (HPLC) | |
| Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-citrulline |
| White-Yellow | |
| MFCD00151943 | |
| fmoc-cit-oh, fmoc-l-citrulline, nalpha-fmoc-l-citrulline, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-ureidopentanoic acid, l-ornithine, n5-aminocarbonyl-n2-9h-fluoren-9-ylmethoxy carbonyl, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-citrulline, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-ureido-pentanoic acid, 2s-5-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-citoh, pubchem19002 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCNC(=O)N)C(=O)O | |
| 397.431 | |
| 397.43 | |
| Crystalline Powder |
Safety and Handling
TSCA : No