missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-citrulline 98.0+%, TCI America™

Supplier:  TCI America F06265G

 View more versions of this product

Catalog No. F06265G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-citrulline
White-Yellow
MFCD00151943
fmoc-cit-oh, fmoc-l-citrulline, nalpha-fmoc-l-citrulline, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-ureidopentanoic acid, l-ornithine, n5-aminocarbonyl-n2-9h-fluoren-9-ylmethoxy carbonyl, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-citrulline, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-ureido-pentanoic acid, 2s-5-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-citoh, pubchem19002
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCNC(=O)N)C(=O)O
397.431
397.43
Crystalline Powder
133174-15-9
C21H23N3O5
5 g
NBMSMZSRTIOFOK-SFHVURJKSA-N
(2S)-5-(carbamoylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
2756127
≥98.0% (HPLC)

Chemical Identifiers

133174-15-9
397.431
NBMSMZSRTIOFOK-SFHVURJKSA-N
2756127
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCNC(=O)N)C(=O)O
C21H23N3O5
MFCD00151943
fmoc-cit-oh, fmoc-l-citrulline, nalpha-fmoc-l-citrulline, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-ureidopentanoic acid, l-ornithine, n5-aminocarbonyl-n2-9h-fluoren-9-ylmethoxy carbonyl, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-citrulline, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-ureido-pentanoic acid, 2s-5-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-citoh, pubchem19002
(2S)-5-(carbamoylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions