Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Wash Bottles

Pour without sputter with these wide-mouth LDPE wash bottles which feature through-the-top closure and stem molded in one piece.
$81.00 - $131.41
Specifications
| Color | Translucent |
|---|---|
| Description | Polypropylene copolymer draw tube |
| Material | LDPE |
| Closure Included | Yes |
| Graduated | No |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
0340915C
![]() |
Thermo Scientific™
24070500 |
500 mL | 53 mm |
Pack of 4 Each for $131.41
Case of 6 Pack for $788.46
|
|
||||
0340915D
![]() |
Thermo Scientific™
24071000 |
1000 mL | 63 mm |
Pack of 2 Each for $81.00
Case of 6 Pack for $486.00
|
|
||||
Description
High quality squeeze bottles with wide mouth for easy filling. Thermo Scientific™ Nalgene™ Wide Mouth LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
- Polypropylene screw closure/stem with PPCO draw tube
- Tip can be cut back for increased flow
- Tight fit of draw tube and closure prevents separation when squeezing bottle
- Flexible LDPE bottle provides soft, ergonomic squeeze
- Sputter-free, precision stream
- Bottle and Carton are readily recyclable in most communities
- Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
| Translucent | |
| LDPE | |
| No | |
| Cylindrical | |
| Polypropylene | |
| Yes |
| Polypropylene copolymer draw tube | |
| Yes | |
| Wide | |
| Wash Bottle | |
| Screw Cap |