Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Bottles with Closure
GREENER_CHOICE
View liquid levels clearly in these impact-resistant, wide-mouth LDPE bottles with closure.
$98.48 - $245.03
Specifications
Color Translucent
Description Wide-Mouth Bottle
Material LDPE
Sterility Non-sterile
Closure Color Natural, translucent
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
028934F
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21030001
30 mL 28 mm
Pack of 12 Each for $98.48
Case of 6 Pack for $590.88
Only null left
028934A
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21030002
60 mL 28 mm
Pack of 12 Each for $100.90
Case of 6 Pack for $605.40
Only null left
028934B
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21030004
125 mL 38 mm
Pack of 12 Each for $127.86
Case of 6 Pack for $767.16
Only null left
028934C
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21030008
250 mL 43 mm
Pack of 12 Each for $182.59
Case of 6 Pack for $1,095.54
Only null left
028934D
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21030016
500 mL 53 mm
Pack of 12 Each for $245.03
Case of 4 Pack for $980.12
Only null left
028934E
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21030032
1 L 63 mm
Pack of 6 Each for $226.31
Case of 4 Pack for $905.24
Only null left
Description
Description
View liquid levels clearly in the impact-resistant Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Bottles with Closure. These translucent, low-density polyethylene bottles are ideal for storing chemicals and standards, as well as collecting and storing distilled water. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. Extremely low trace metal content in LDPE makes it ideal for trace metal analysis. Excellent chemical resistance to most acids, bases and alcohols. Wide mouth is easy to fill. Leakproof† when used with Nalgene closures.
  • Extremely low trace metal content in LDPE makes it ideal for trace metal analysis
  • Excellent chemical resistance to most acids, bases and alcohols make this a versatile bottle; suitable for shipping liquids
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Linerless caps ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate solutions
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Includes:
Polypropylene screw caps
Note: These tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof† when used with their Nalgene closures.
.
Recycling Information:
Nalgene reusable LDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
Specifications
Translucent
LDPE
Natural, translucent
Yes
Round
Unlined
Screw Cap
Wide-Mouth Bottle
Non-sterile
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Wide
Wide Mouth Bottle
Polypropylene
No
Videos
SDS
Documents
Documents
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.