Learn More
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing, and storage.
$360.34 - $1139.44
Specifications
| Material | PETG |
|---|---|
| No. per Pack | 1 |
| Shape | Erlenmeyer |
| Surface Coating | None |
| Autoclavable | Not Autoclavable |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Closure Type | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Closure Type | Price | Quantity | |||
|
10-524
|
Thermo Scientific™
41150125 |
125 mL | 38-430 mm | Vented |
Case of 24 Each for $1,139.44
|
|
|||
|
10-525
|
Thermo Scientific™
41150250 |
250 mL | 38-430 mm | Vented |
Case of 12 Each for $667.94
|
|
|||
|
10-526
|
Thermo Scientific™
41150500 |
500 mL | 45-430 mm | Vented |
Case of 12 Each for $838.22
|
|
|||
|
10-527
|
Thermo Scientific™
41151000 |
1000 mL | 45-430 mm | Vented |
Case of 6 Each for $525.72
|
|
|||
|
10-528
|
Thermo Scientific™
41152000 |
2000 mL | 45-430 mm | Vented |
Case of 4 Each for $455.13
|
|
|||
|
03-600-152
|
Thermo Scientific™
41152800 |
2800 mL | 70 mm | Vented |
Case of 4 Each for $755.16
|
|
|||
|
10-041-20
|
Thermo Scientific™
41120125 |
125 mL | 38-430 mm | Solid, non-vented |
Case of 24 Each for $926.63
|
|
|||
|
10-346-7A
|
Thermo Scientific™
41120250 |
250 mL | 38-430 mm | Solid, non-vented |
Case of 12 Each for $573.00
|
|
|||
|
10-346-7B
|
Thermo Scientific™
41120500 |
500 mL | 45-430 mm | Solid, non-vented |
Case of 12 Each for $723.63
|
|
|||
|
10-041-21
|
Thermo Scientific™
41121000 |
1000 mL | 45-430 mm | Solid, non-vented |
Case of 6 Each for $476.53
|
|
|||
|
10-346-7C
|
Thermo Scientific™
41122000 |
2000 mL | 45-430 mm | Solid, non-vented |
Case of 4 Each for $360.34
|
|
|||
|
03-600-150
|
Thermo Scientific™
41122800 |
2800 mL | 70 mm | Solid, non-vented |
Case of 4 Each for $700.81
|
|
|||
Description
Save preparation time and avoid contamination risk with Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing, and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Features
- Sterile and single use for reduced contamination risk
- BPA-free
- Collapses when autoclaved, reducing biohazardous waste
- Graduated for quick volume assessment
- Regulatory documentation available on request
Warranty: 90 days
Specifications
| PETG | |
| Erlenmeyer | |
| Not Autoclavable | |
| Yes | |
| Plain |
| 1 | |
| None | |
| Single-use | |
| Sterile |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.