missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Replacement Silicone O-Ring for Centrifuge Bottle Sealing Cap

Description
The Nalgene™ Replacement Silicone O-Ring is used as a replacement for the sealing cap on Nalgene™ Cat #3141-1006 Super-Speed Centrifuge Bottle.
- Replacement O-ring
- Used for cap of Nalgene™ Super-Speed Centrifuge Bottle #3141-1006
- Silicone
- 85mm size
- Red
Specifications
Specifications
| Color | Red |
| Material | Silicone |
| For Use With (Equipment) | Nalgene Cat #3141-1006 Super-Speed Centrifuge Bottle Cap |
| Closure Type | Sealing Cap |
| Shape | Round |
| Product Type | O-ring |