missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
GREENER_CHOICE

Catalog No. p-4521570
Change view
Click to view available options
Nominal Capacity:
250 mL
450 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Nominal Capacity
055641 250 mL
055643 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 055641 Supplier Thermo Scientific™ Supplier No. 31410250
Only null left

Centrifugalize low- to moderate-speed, large-volume, biological and chemical samples with these PPCO centrifuge bottles, which come with a sealing closure.

Centrifugalize low- to moderate-speed, large-volume, biological and chemical samples with Thermo Scientific™ Nalgene™ PPCO Centrifuge Bottles, which include a sealing closure for added leakproof† assurance at maximum speeds when spinning hazardous materials. Molded of rigid and robust polypropylene copolymer, these translucent bottles are designed for pelleting applications, such as cell culture harvesting and protein biochemistry, including ammonium sulfate precipitations. Additionally, these PPCO bottles are autoclavable and offer excellent mechanical strength.

Features:
  • For low- to moderate-speed centrifugation of biological and chemical samples
  • Bottle includes polypropylene sealing cap assembly with silicone gasket for added leakproof† assurance at maximum speeds or when spinning hazardous materials
  • For proper performance, bottles must be filled to 80% of total capacity

Requires:

When spinning 250mL bottle in a Sorvall GSA rotor, use of the Nalgene centrifuge bottle adapter (DS3125-0250) is required

Specifications

Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Translucent
Diameter (Metric) Outer Neck 60.7 mm
Material PPCO
Length (Metric) 133.4 mm
No. per Pack 4
Nominal Capacity 250 mL
Closure Included Yes
Graduated No
Max. Force 27,500 x g
Shape Round
Height (Metric) 132.8 mm
Product Type Centrifuge Bottle
Width (English) 2.40 in.
Length (English) 5.23 in.
Height (English) 5.23 in.
Closure Material Polypropylene, red rubber
Diameter (Metric) 61.1 mm
Closure Type Sealing Cap Assembly with Silicone Gasket
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less