missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels
GREENER_CHOICE

Catalog No. 10348C
Change view
Click to view available options
Diameter (Metric)Top:
64 mm
79mm
100 mm
145 mm
Capacity (Metric):
68 mL
115 mL
243 mL
716 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)Top Capacity (Metric)
10348C 100 mm 243 mL
10348A 64 mm 68 mL
10348B 79mm 115 mL
10348D 145 mm 716 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10348C Supplier Thermo Scientific™ Supplier No. 42520100
Edge

Transfer powders with these powder funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. A shatter-proof alternative to glass.

Transfer powders with Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.

  • Polypropylene is break-resistant and chemically compatible with a wide variety of lab reagents
  • Suitable for general lab use, it is a durable replacement for glass funnels
  • Autoclavable

Specifications

Diameter (Metric)Top 100 mm
Capacity (Metric) 243 mL
Diameter (Metric) Stem 23 mm
Material Polypropylene
Height (Metric) 105 mm
Autoclavable Autoclavable
Description Funnel Powder PP 100mm
Length (Metric) Stem 34 mm
Quantity 6/Pk.; 24/Cs.