missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polycarbonate Centrifuge Bottles
GREENER_CHOICE

Catalog No. p-4521563
Change view
Click to view available options
Nominal Capacity:
500 mL
1000 mL
For Use With (Equipment):
Fiberlite™ rotors
IEC rotors
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Nominal Capacity For Use With (Equipment)
0557924 500 mL Fiberlite™ rotors
0557925 1000 mL Fiberlite™ rotors
0557926 1000 mL IEC rotors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0557924 Supplier Thermo Scientific™ Supplier No. 31220500
Only null left

Process large-volume cell harvesting, pelleting and protein purification with these polycarbonate centrifuge bottles.

Process large-volume cell harvesting, pelleting and protein purification with Thermo Scientific™ Nalgene™ Polycarbonate Centrifuge Bottles, the perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors. Molded of transparent polycarbonate, these clear bottles are autoclavable and have excellent mechanical strength properties.

Molded of transparent polycarbonate for easy visualization of samples, these bottles are ideal for low-speed centrifugation of biological and chemical samples.

  • For added leakproof† assurance at maximum speeds or when spinning hazardous materials, use with polypropylene sealing caps assemblies; available for 250mL (DS3131-0038) and 1000mL (DS3132-0063) sizes
  • Cat. No. 3122-1010 designed for use with IEC rotors
  • For proper performance, bottles must be filled to at least 80% of total capacity

Ordering Information:
When spinning a 250mL bottle in a Thermo Scientific Sorvall GSA rotor, the use of the Nalgene centrifuge bottle adaptor (DS3125-0250) is required. Cat. No. 3122-1010 is for use with IEC Rotors.

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Color Clear
Description Polycarbonate Centrifuge Bottles
Material Polycarbonate
No. per Case 24
No. per Pack 4
Nominal Capacity 500 mL
Closure Included Yes
Graduated No
Max. Force 13,700 x g
Shape Round
Height (Metric) 168.9 mm
Product Type Centrifuge Bottle
For Use With (Equipment) Fiberlite™ rotors
Closure Material Polypropylene
Diameter (Metric) 69.5 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.