missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polycarbonate Centrifuge Bottles with Sealing Closure
GREENER_CHOICE

Catalog No. 05564
Change view
Click to view available options
Nominal Capacity:
250 mL
450 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Nominal Capacity
05564 250 mL
055642 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05564 Supplier Thermo Scientific™ Supplier No. 31400250
Get a Water Bottle with Plastics Purchase
View all promotions

Process large-volume cell harvesting, pelleting and protein purification with these polycarbonate centrifuge bottles.

Ideal for large-volume processes such as cell harvesting, pelleting, and protein purification, Thermo Scientific™ Nalgene™ PC Centrifuge Bottles are molded of crystal-clear polycarbonate, which allows easy sample visualization and results in excellent mechanical strength. These centrifuge bottles are the perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors.

  • Molded of polycarbonate for easy visualization of samples, these bottles are ideal for low- to moderate-speed centrifugation of biological and chemical samples
  • Polypropylene sealing cap closure with silicone gasket adds leakproof† assurance at maximum speeds or when spinning hazardous materials
  • For proper performance, bottles must be filled to 80% of total capacity

Ordering Information:
When spinning 250mL bottle in a Thermo Scientific™ Sorvall™ GSA rotor, use of the Nalgene centrifuge bottle adapter (DS3125-0250) is required.

Specifications

Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Clear
Material Polycarbonate
No. per Case 36
No. per Pack 4
Nominal Capacity 250 mL
Closure Included Yes
Graduated No
Max. Force 27,500 x g
Shape Round
Height (Metric) 134.4 mm
Product Type Centrifuge Bottle
Width (English) 2.432 in.
Length (English) 2.432 in.
Height (English) 5.29 in.
Closure Material Polypropylene, red rubber
Diameter (Metric) 61.5 mm
Closure Type Sealing Cap w/Silicone Gasket
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.