missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays

Pack and ship liquid medias, buffers and sera using these heavy-walled, durable PETG square media bottles, designed for use in filling lines.
Specifications
| Color | Clear |
|---|---|
| Description | Media bottles without closure |
| For Use With (Application) | Filling Lines |
| Material | PETG |
| Sterility | Sterile |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
0331389D
![]() |
Thermo Scientific™
3420240250 |
250 mL | 38-430 mm |
N/A
|
|
||||
0331389E
![]() |
Thermo Scientific™
3420240500 |
500 mL | 38-430 mm |
N/A
|
|
||||
0331389G
![]() |
Thermo Scientific™
3420241000 |
1 L | 38-430 mm |
N/A
|
|
||||
Description
Pack and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles are designed for use in filling lines; closures sold separately.
Features:- Heavy-walled and durable
- Shatter-resistant
- Excellent gas barrier properties
- Sterile 10-6 SAL
- Molded in graduations
- Closure supplied separately
Specifications
| Clear | |
| Filling Lines | |
| Sterile | |
| Yes | |
| Square | |
| Media Bottle |
| Media bottles without closure | |
| PETG | |
| No | |
| Narrow | |
| Sterile |