missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
GREENER_CHOICE

Catalog No. 0331389G
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
Closure Size:
38-430 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
0331389G 1 L 38-430 mm
0331389D 250 mL 38-430 mm
0331389E 500 mL 38-430 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0331389G Supplier Thermo Scientific™ Supplier No. 3420241000

Pack and ship liquid medias, buffers and sera using these heavy-walled, durable PETG square media bottles, designed for use in filling lines.

Pack and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles are designed for use in filling lines; closures sold separately.

Features:
  • Heavy-walled and durable
  • Shatter-resistant
  • Excellent gas barrier properties
  • Sterile

    10-6

    SAL
  • Molded in graduations
  • Closure supplied separately

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1 L
Capacity (English) 32 oz.
Color Clear
Description Media bottles without closure
Diameter (English) 3.6 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
For Use With (Application) Filling Lines
Material PETG
No. per Case 24
No. per Pack 12
Sterility Sterile
Closure Included No
Graduated Yes
Mouth Narrow
Shape Square
Fit for Purpose Sterile
Height (Metric) 220 mm
Product Type Media Bottle
Height (English) 8.66 in.
Closure Size 38-430 mm
Diameter (Metric) 92 mm
Show More Show Less