Learn More
Thermo Scientific™ Nalgene™ PETG Low Particulate Media Bottles
Nalgene PETG Low Particulate Media Bottles are certified clean to USP <788> standards, making them an ideal solution for biological and pharmaceutical manufacturing.
$1606.23 - $2633.82
Specifications
| For Use With (Application) | Media/Buffer Preparation â, Media/Buffer Storage, Sampling, Storage, Transportation â, Validation Testing, Drug Substance, Drug Product |
|---|---|
| Material | PETG |
| Sterility | Sterile |
| Temperature Range (Metric) | -70°C to 70°C |
| Low Particulate | USP <788> (Low particulate) |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
15-350-201
|
Thermo Scientific™
3820190030 |
30 mL |
Case of 72 Each for $1,606.23
|
|
|||||
|
15-350-202
|
Thermo Scientific™
3820190060 |
60 mL |
Case of 72 Each for $2,261.07
|
|
|||||
|
15-350-203
|
Thermo Scientific™
3820190125 |
125 mL |
Case of 48 Each for $2,305.21
|
|
|||||
|
15-350-204
|
Thermo Scientific™
3820190250 |
250 mL |
Case of 48 for $2,633.82
|
|
|||||
|
15-350-205
|
Thermo Scientific™
3820190500 |
500 mL |
Case of 24 Each for $1,743.82
|
|
|||||
|
15-350-207
|
Thermo Scientific™
3820192000 |
2 L |
Case of 12 for $1,957.68
|
|
|||||
|
15-350-206
|
Thermo Scientific™
3820191000 |
1 L |
Case of 24 for $2,445.39
|
|
|||||
Description
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific™ Nalgene™ PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.
Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.
- Exhibit excellent gas barrier properties and ensure pH stability for buffered solutions.
- Resins are free of animal-derived components.
- Sterile, tamper-resistant seal ensures product integrity prior to use.
Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
| Media/Buffer Preparation â, Media/Buffer Storage, Sampling, Storage, Transportation â, Validation Testing, Drug Substance, Drug Product | |
| Sterile | |
| USP <788> (Low particulate) | |
| Yes | |
| Narrow | |
| Clean Container | |
| Upstream media/buffer prep and storage. Upstream process solution storage. Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation. |
| PETG | |
| -70°C to 70°C | |
| USP Class VI, European Pharmacopeia for Abnormal Toxicity and USP 661 physicochemical tests compliant. Manufactured in an ISO 14644-1 Class 7 cleanroom. Certified sterile (SAL 10-6), non-cytotoxic, non-pyrogenic, and non-hemolytic. | |
| Yes | |
| Square | |
| Not Autoclavable |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.