missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Low Particulate Media Bottles
GREENER_CHOICE

Catalog No. 15350201
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
2 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
15350201 30 mL
15350202 60 mL
15350203 125 mL
15350204 250 mL
15350205 500 mL
15350207 2 L
15350206 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 15350201 Supplier Thermo Scientific™ Supplier No. 3820190030

Nalgene PETG Low Particulate Media Bottles are certified clean to USP <788> standards, making them an ideal solution for biological and pharmaceutical manufacturing.

Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific™ Nalgene™ PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.

Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.

  • Exhibit excellent gas barrier properties and ensure pH stability for buffered solutions.
  • Resins are free of animal-derived components.
  • Sterile, tamper-resistant seal ensures product integrity prior to use.

Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Diameter (English) 1.5 in.
Diameter (English) Inner Neck 0.55 in.
Diameter (Metric) Inner Neck 14 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Material PETG
No. per Case 72/Cs.
No. per Pack 6/Pk.
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
Low Particulate USP <788> (Low particulate)
Certifications/Compliance USP Class VI, European Pharmacopeia for Abnormal Toxicity and USP 661 physicochemical tests compliant. Manufactured in an ISO 14644-1 Class 7 cleanroom. Certified sterile (SAL 10-6), non-cytotoxic, non-pyrogenic, and non-hemolytic.
Closure Included Yes
Graduated Yes
Mouth Narrow
Shape Square
Height (Metric) 64 mm
Product Type Clean Container
Height (English) 2.52 in.
Closure Size 20 mm
Diameter (Metric) 38 mm
Autoclavable Not Autoclavable
Workflow Step Upstream media/buffer prep and storage. Upstream process solution storage. Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation.
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.