missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Low Particulate Media Bottles

Nalgene PETG Low Particulate Media Bottles are certified clean to USP <788> standards, making them an ideal solution for biological and pharmaceutical manufacturing.

Supplier:  Thermo Scientific™ 3820190500

 View more versions of this product

Catalog No. 15-350-205


Only null left
Explore available promotions

Description

Description

Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific™ Nalgene™ PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.

Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.

  • Exhibit excellent gas barrier properties and ensure pH stability for buffered solutions.
  • Resins are free of animal-derived components.
  • Sterile, tamper-resistant seal ensures product integrity prior to use.

Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications

Specifications

500 mL
2.9 in.
28 mm
PETG
4/Pk.
-70°C to 70°C
USP Class VI, European Pharmacopeia for Abnormal Toxicity and USP 661 physicochemical tests compliant. Manufactured in an ISO 14644-1 Class 7 cleanroom. Certified sterile (SAL 10-6), non-cytotoxic, non-pyrogenic, and non-hemolytic.
Yes
Square
Clean Container
38-430 mm
Not Autoclavable
16.9 oz.
1.1 in.
Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
24/Cs.
Sterile
USP <788> (Low particulate)
Yes
Narrow
177 mm
6.97 in.
74 mm
Upstream media/buffer prep and storage. Upstream process solution storage. Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.