Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Bottles with Closure: Autoclavable

Perform lab applications that require excellent chemical resistance and autoclaving, with and without contents, using this polypropylene copolymer bottle with closure.
$54.41 - $461.62
Specifications
| Color | Translucent |
|---|---|
| Description | Autoclavable Narrow-Mouth Bottle |
| Material | PPCO |
| Sterility | Non-sterile |
| Certifications/Compliance | Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. |
Alert:
Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
0292311E
![]() |
Thermo Scientific™
20069125 |
4 mL | 13 mm |
Pack of 12 Each for $54.41
Case of 6 Pack for $326.46
|
|
||||
0292311D
![]() |
Thermo Scientific™
20069025 |
8 mL | 20 mm |
Pack of 12 Each for $56.31
Case of 6 Pack for $337.86
|
|
||||
029238G
![]() |
Thermo Scientific™
20069050 |
15 mL | 20 mm |
Pack of 12 Each for $62.97
Case of 6 Pack for $377.82
|
|
||||
02925A
![]() |
Thermo Scientific™
20060001 |
30 mL | 20 mm |
Case of 72 Each for $461.62
|
|
||||
02925B
![]() |
Thermo Scientific™
20060002 |
60 mL | 20 mm |
Pack of 12 Each for $79.76
Case of 6 Pack for $478.56
|
|
||||
02925C
![]() |
Thermo Scientific™
20060004 |
125 mL | 24 mm |
Pack of 12 Each for $107.90
Case of 6 Pack for $647.40
|
|
||||
02925D
![]() |
Thermo Scientific™
20060008 |
250 mL | 24 mm |
Pack of 12 Each for $157.17
Case of 6 Pack for $943.02
|
|
||||
02925E
![]() |
Thermo Scientific™
20060016 |
500 mL | 28 mm |
Pack of 12 Each for $232.93
Case of 4 Pack for $931.72
|
|
||||
02925F
![]() |
Thermo Scientific™
20060032 |
1 L | 38-430 mm |
Pack of 6 Each for $216.52
Case of 4 Pack for $866.08
|
|
||||
Description
Perform lab applications requiring excellent chemical resistance and autoclaving, with and without contents, using Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Bottles with Closure. These polypropylene copolymer bottles are translucent, providing good clarity for viewing contents. Made only from high-quality, laboratory-grade plastic materials.
- Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
- Autoclavable
- Leakproof† when used with Nalgene closures
- Bottle and carton are readily recyclable in most communities
- Manufactured from non-toxic materials
Note:
Completely disengage threads or remove cap before autoclaving.
Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.Recycling Information:
Nalgene reusable polypropylene (PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.
Specifications
| Translucent | |
| PPCO | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
| Narrow | |
| Narrow Mouth Bottle | |
| Polypropylene | |
| No |
| Autoclavable Narrow-Mouth Bottle | |
| Non-sterile | |
| Yes | |
| Round | |
| Unlined | |
| Screw Cap | |
| Autoclavable |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.