Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Bottles with Closure: Autoclavable

Perform lab applications that require excellent chemical resistance and autoclaving, with and without contents, using this polypropylene copolymer bottle with closure.
Supplier: Thermo Scientific™ 20060032
Alert:
Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.
Description
Perform lab applications requiring excellent chemical resistance and autoclaving, with and without contents, using Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Bottles with Closure. These polypropylene copolymer bottles are translucent, providing good clarity for viewing contents. Made only from high-quality, laboratory-grade plastic materials.
- Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
- Autoclavable
Note:
Completely disengage threads or remove cap before autoclaving.
Chemical Identifiers
| Translucent | |
| PPCO | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
| Narrow | |
| Narrow Mouth Bottle | |
| Polypropylene | |
| No |
| Autoclavable Narrow-Mouth Bottle | |
| Non-sterile | |
| Yes | |
| Round | |
| Unlined | |
| Screw Cap | |
| Autoclavable |
Specifications
| 32 oz., 1000 mL | |
| 32 oz. | |
| Autoclavable Narrow-Mouth Bottle | |
| PPCO | |
| 6 | |
| Natural | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
| Reusable | |
| Round | |
| Narrow Mouth Bottle | |
| Unlined | |
| 38-430 mm | |
| Screw Cap | |
| Autoclavable |
| 1 L | |
| Translucent | |
| 27 mm | |
| 24 | |
| Non-sterile | |
| 38 mm | |
| Yes | |
| Narrow | |
| 216 mm | |
| 8.5 in. | |
| Polypropylene | |
| 91 mm | |
| No |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.