Learn More
Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles
GREENER_CHOICE
Prevent separation when squeezing these LDPE economy wash bottles, which feature a tight-fitting draw tube and closure.
$101.10 - $147.62
Specifications
Color Translucent
Includes Stem and draw tube
Material LDPE
Closure Included Yes
Graduated No
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
0340910AA
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
24010125
125 mL 24 mm
Pack of 6 Each for $101.10
Case of 8 Pack for $808.80
Only null left
0340910BB
GREENER_CHOICE
Accessories View Documents Promotion Details
Thermo Scientific™
24010250
250 mL 24 mm
Pack of 6 Each for $115.55
Case of 6 Pack for $693.30
Only null left
0340910CC
GREENER_CHOICE
Accessories View Documents Promotion Details
Thermo Scientific™
24010500
500 mL 28 mm
Pack of 6 Each for $147.62
Case of 4 Pack for $590.48
Only null left
0340910DD
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
24011000
1000 mL 38-430 mm
Pack of 4 Each for $117.48
Case of 3 Pack for $352.44
Only null left
Description
Description
High quality squeeze bottles at a value price. Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
Features:
  • Polypropylene screw closure/stem with PPCO draw tube
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Sputter-free, precision stream
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
Specifications
Translucent
LDPE
No
Cylindrical
Polypropylene
Yes
Stem and draw tube
Yes
Narrow
Economy Wash Bottle
Screw Cap