missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles
GREENER_CHOICE

Catalog No. 0340910AA
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Closure Size:
24 mm
28 mm
38-430 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
0340910AA 125 mL 24 mm
0340910BB 250 mL 24 mm
0340910CC 500 mL 28 mm
0340910DD 1000 mL 38-430 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0340910AA Supplier Thermo Scientific™ Supplier No. 24010125
Get a Water Bottle with Plastics Purchase
View all promotions

Prevent separation when squeezing these LDPE economy wash bottles, which feature a tight-fitting draw tube and closure.

High quality squeeze bottles at a value price. Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.

Features:

  • Polypropylene screw closure/stem with PPCO draw tube
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle

Specifications

Capacity 4 oz., 125 mL
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Translucent
Includes Stem and draw tube
Material LDPE
No. per Case 48
No. per Pack 6
Closure Included Yes
Graduated No
Mouth Narrow
Shape Cylindrical
Height (Metric) 147 mm
Product Type Economy Wash Bottle
Height (English) 5.8 in.
Closure Material Polypropylene
Closure Size 24 mm
Diameter (Metric) 50.3 mm
Closure Type Screw Cap
Through the Closure Dispensing Yes
Show More Show Less