missing translation for 'onlineSavingsMsg'
Learn More

Nafamostat Mesylate 98.0+%, TCI America™

Catalog No. p-7134049
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N095920MG 20 mg
N0959100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N095920MG Supplier TCI America Supplier No. N095920MG

Chemical Identifiers

CAS 82956-11-4
Molecular Formula C21H25N5O8S2
Molecular Weight (g/mol) 539.58
MDL Number MFCD00941430
InChI Key SRXKIZXIRHMPFW-UHFFFAOYSA-N
Synonym nafamostat mesylate, futhan, nafamostat mesilate, nafamstat mesilate, 6-amidino-2-naphthyl 4-guanidino-benzoate dimethanesulphonate, 6-amidino-2-naphthyl-4-guanidinobenzoate dimethanesulfonate, nafamostat mesylate fut-175, nafamstatmesilate, ronastat tn
PubChem CID 5311180
IUPAC Name amino({[4-({[6-(azaniumylmethanimidoyl)naphthalen-2-yl]oxy}carbonyl)phenyl]imino})methanaminium dimethanesulfonate
SMILES CS([O-])(=O)=O.CS([O-])(=O)=O.NC([NH3+])=NC1=CC=C(C=C1)C(=O)OC1=CC=C2C=C(C=CC2=C1)C([NH3+])=N

Specifications

Melting Point 258°C
Color White
Quantity 20 mg
Formula Weight 539.58
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nafamostat Mesylate
RTECSNumber DG2736000
TSCA No