accessibility menu, dialog, popup

UserName

CH3SOOOCH3SOOOH2NNH3NOONH3NH(E/Z)

CAS RN 82956-11-4

IUPAC Name: amino({[4-({[6-(azaniumylmethanimidoyl)naphthalen-2-yl]oxy}carbonyl)phenyl]imino})methanaminium dimethanesulfonate
Synonyms: amino({[4-({[6-(ammoniomethanimidoyl)naphthalen-2-yl]oxy}carbonyl)phenyl]imino})methanaminium dimesylate
Molecular Weight (g/mol): 539.58
Molecular Formula: C21H25N5O8S2
InChi Key: SRXKIZXIRHMPFW-UHFFFAOYSA-N
SMILES: CS([O-])(=O)=O.CS([O-])(=O)=O.NC([NH3+])=NC1=CC=C(C=C1)C(=O)OC1=CC=C2C=C(C=CC2=C1)C([NH3+])=N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results