missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NADA-green

Catalog No. 66485
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66485 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66485 Supplier Tocris Bioscience™ Supplier No. 6648/5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fluorescent Probes for Imaging Bacteria

NADA-green is a fluorescent D-amino acid. Suitable for labeling peptidoglycans in live bacteria. Incorporated into bacterial cell walls during synthesis. Results in strong peripheral and septal labeling of taxonomically diverse bacterial cell populations without affecting growth rate. Excitation/emission λ ~450/555 nm.

please note that the spectral data is solvent dependant

Specifications

Chemical Name or Material 3-[(7-Nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-alanine hydrochloride
CAS 2253733-11-6
Quantity 5 mg
Molecular Formula C9H9N5O5.HCl
Formula Weight 303.66
Purity 98%