missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nabumetone, MilliporeSigma™ Supelco™
$273.24 - $472.39
Specifications
| CAS | 42924-53-8 |
|---|---|
| Physical Form | Neat |
| Linear Formula | C15H16O2 |
| MDL Number | MFCD00079518 |
| Synonym | 4-(6-Methoxy-2-naphthyl)-2-butanone |
Description
Featured Industries: Pharmaceutical (small molecule)Specifications
| 42924-53-8 | |
| C15H16O2 | |
| 4-(6-Methoxy-2-naphthyl)-2-butanone | |
| 228.29 |
| Neat | |
| MFCD00079518 | |
| Analytical Standard | |
| Analytical Standard |