missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N6-Methyl-2'-deoxyadenosine, 99%
CAS: 2002-35-9 | C11H15N5O3 | 265.27 g/mol
$264.63 - $1,008.01
Chemical Identifiers
| CAS | 2002-35-9 |
|---|---|
| Molecular Formula | C11H15N5O3 |
| Molecular Weight (g/mol) | 265.27 |
| MDL Number | MFCD00055999 |
| InChI Key | DYSDOYRQWBDGQQ-XLPZGREQSA-N |
| PubChem CID | 168948 |
| ChEBI | CHEBI:7417 |
| SMILES | CNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ64961MD
|
Thermo Scientific Chemicals
J64961MD |
250 mg |
Each for $264.63
|
|
|||||
|
AAJ6496103
|
Thermo Scientific Chemicals
J6496103 |
1 g |
Each for $1,008.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2002-35-9 | |
| 265.27 | |
| DYSDOYRQWBDGQQ-XLPZGREQSA-N | |
| CHEBI:7417 |
| C11H15N5O3 | |
| MFCD00055999 | |
| 168948 | |
| CNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 |
Specifications
| 2002-35-9 | |
| C11H15N5O3 | |
| n6-methyl-2'-deoxyadenosine, n6-me-dado, n 6-me-dado, adenosine, 2'-deoxy-n-methyl, unii-48qzw2ir9h, 2r,3s,5r-2-hydroxymethyl-5-6-methylamino-9h-purin-9-yl tetrahydrofuran-3-ol, n 6-methyl-2'-deoxyadenosine, n6-methyldeoxyadenosine, 2'-deoxy-n6-methylado, 48qzw2ir9h | |
| DYSDOYRQWBDGQQ-XLPZGREQSA-N | |
| (2R,3S,5R)-2-(hydroxymethyl)-5-[6-(methylamino)-9H-purin-9-yl]oxolan-3-ol | |
| 168948 | |
| 265.27 | |
| Powder |
| White | |
| MFCD00055999 | |
| Soluble in water | |
| CNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 | |
| 265.27 | |
| CHEBI:7417 | |
| 99% | |
| N6-Methyl-2'-deoxyadenosine |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only