missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N6-Methyl-2'-deoxyadenosine, 99%

Catalog No. AAJ64961MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64961MD 250 mg
AAJ6496103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64961MD Supplier Thermo Scientific Chemicals Supplier No. J64961MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2002-35-9 | C11H15N5O3 | 265.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2002-35-9
Molecular Formula C11H15N5O3
Molecular Weight (g/mol) 265.27
MDL Number MFCD00055999
InChI Key DYSDOYRQWBDGQQ-XLPZGREQSA-N
PubChem CID 168948
ChEBI CHEBI:7417
SMILES CNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1

Specifications

Color White
Quantity 250 mg
Synonym n6-methyl-2'-deoxyadenosine, n6-me-dado, n 6-me-dado, adenosine, 2'-deoxy-n-methyl, unii-48qzw2ir9h, 2r,3s,5r-2-hydroxymethyl-5-6-methylamino-9h-purin-9-yl tetrahydrofuran-3-ol, n 6-methyl-2'-deoxyadenosine, n6-methyldeoxyadenosine, 2'-deoxy-n6-methylado, 48qzw2ir9h
Solubility Information Soluble in water
IUPAC Name (2R,3S,5R)-2-(hydroxymethyl)-5-[6-(methylamino)-9H-purin-9-yl]oxolan-3-ol
Formula Weight 265.27
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N6-Methyl-2'-deoxyadenosine
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only