missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N6-Methyl-2'-deoxyadenosine, 99%

CAS: 2002-35-9 | C11H15N5O3 | 265.27 g/mol

Supplier:  Thermo Scientific Chemicals J6496103

Quantity 1 g

 View more versions of this product

Catalog No. AAJ6496103


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2002-35-9
265.27
DYSDOYRQWBDGQQ-XLPZGREQSA-N
CHEBI:7417
C11H15N5O3
MFCD00055999
168948
CNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1

Specifications

N6-Methyl-2'-deoxyadenosine
White
1 g
n6-methyl-2'-deoxyadenosine, n6-me-dado, n 6-me-dado, adenosine, 2'-deoxy-n-methyl, unii-48qzw2ir9h, 2r,3s,5r-2-hydroxymethyl-5-6-methylamino-9h-purin-9-yl tetrahydrofuran-3-ol, n 6-methyl-2'-deoxyadenosine, n6-methyldeoxyadenosine, 2'-deoxy-n6-methylado, 48qzw2ir9h
DYSDOYRQWBDGQQ-XLPZGREQSA-N
(2R,3S,5R)-2-(hydroxymethyl)-5-[6-(methylamino)purin-9-yl]oxolan-3-ol
168948
265.27
Powder
2002-35-9
C11H15N5O3
MFCD00055999
Soluble in water
CNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
265.27
CHEBI:7417
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions