missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-Dibenzoyladenosine 2',3'-Dibenzoate 98.0+%, TCI America™
$132.90 - $674.02
Chemical Identifiers
| CAS | 58463-04-0 |
|---|---|
| Molecular Formula | C38H29N5O8 |
| Molecular Weight (g/mol) | 683.677 |
| InChI Key | GNYQOUZNLMTKQG-WVPZXSLWSA-N |
| PubChem CID | 44630009 |
| IUPAC Name | [(2R,3R,5R)-4-benzoyloxy-5-[6-(dibenzoylamino)purin-9-yl]-2-(hydroxymethyl)oxolan-3-yl] benzoate |
| SMILES | C1=CC=C(C=C1)C(=O)N(C2=NC=NC3=C2N=CN3C4C(C(C(O4)CO)OC(=O)C5=CC=CC=C5)OC(=O)C6=CC=CC=C6)C(=O)C7=CC=CC=C7 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
B3460100MG
|
N/A | 100 mg |
Each for $132.90
|
|
|||||
|
B34601G
|
N/A | 1 g |
Each for $674.02
|
|
|||||
Chemical Identifiers
| 58463-04-0 | |
| 683.677 | |
| 44630009 | |
| C1=CC=C(C=C1)C(=O)N(C2=NC=NC3=C2N=CN3C4C(C(C(O4)CO)OC(=O)C5=CC=CC=C5)OC(=O)C6=CC=CC=C6)C(=O)C7=CC=CC=C7 |
| C38H29N5O8 | |
| GNYQOUZNLMTKQG-WVPZXSLWSA-N | |
| [(2R,3R,5R)-4-benzoyloxy-5-[6-(dibenzoylamino)purin-9-yl]-2-(hydroxymethyl)oxolan-3-yl] benzoate |
Specifications
| 58463-04-0 | |
| White | |
| GNYQOUZNLMTKQG-WVPZXSLWSA-N | |
| [(2R,3R,5R)-4-benzoyloxy-5-[6-(dibenzoylamino)purin-9-yl]-2-(hydroxymethyl)oxolan-3-yl] benzoate | |
| 44630009 | |
| ≥98.0% (HPLC,T) | |
| N6-Dibenzoyladenosine 2′,3′-Dibenzoate |
| 183°C | |
| C38H29N5O8 | |
| C1=CC=C(C=C1)C(=O)N(C2=NC=NC3=C2N=CN3C4C(C(C(O4)CO)OC(=O)C5=CC=CC=C5)OC(=O)C6=CC=CC=C6)C(=O)C7=CC=CC=C7 | |
| 683.677 | |
| 683.68 | |
| Crystalline Powder |
Safety and Handling
TSCA : No