missing translation for 'onlineSavingsMsg'
Learn More

N6-Dibenzoyladenosine 2',3'-Dibenzoate 98.0+%, TCI America™

Catalog No. B34601G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34601G 1 g
B3460100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34601G Supplier TCI America Supplier No. B34601G

Chemical Identifiers

CAS 58463-04-0
Molecular Formula C38H29N5O8
Molecular Weight (g/mol) 683.677
InChI Key GNYQOUZNLMTKQG-WVPZXSLWSA-N
PubChem CID 44630009
IUPAC Name [(2R,3R,5R)-4-benzoyloxy-5-[6-(dibenzoylamino)purin-9-yl]-2-(hydroxymethyl)oxolan-3-yl] benzoate
SMILES C1=CC=C(C=C1)C(=O)N(C2=NC=NC3=C2N=CN3C4C(C(C(O4)CO)OC(=O)C5=CC=CC=C5)OC(=O)C6=CC=CC=C6)C(=O)C7=CC=CC=C7

Specifications

Melting Point 183°C
Color White
Quantity 1 g
Formula Weight 683.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N6-Dibenzoyladenosine 2′,3′-Dibenzoate