missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-Benzoyladenosine Hydrate 96.0+%, TCI America™
$79.74 - $320.87
Chemical Identifiers
| CAS | 4546-55-8 |
|---|---|
| Molecular Formula | C17H17N5O5 |
| Molecular Weight (g/mol) | 371.35 |
| MDL Number | MFCD00056988 |
| InChI Key | NZDWTKFDAUOODA-UCQOUBKUNA-N |
| PubChem CID | 11728391 |
| IUPAC Name | N-{9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide |
| SMILES | OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12 |
Chemical Identifiers
| 4546-55-8 | |
| 371.35 | |
| NZDWTKFDAUOODA-UCQOUBKUNA-N | |
| N-{9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide |
| C17H17N5O5 | |
| MFCD00056988 | |
| 11728391 | |
| OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12 |
Specifications
| 4546-55-8 | |
| White | |
| MFCD00056988 | |
| NZDWTKFDAUOODA-UCQOUBKUNA-N | |
| N-{9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide | |
| 11728391 | |
| ≥96.0% (HPLC,T) | |
| N6-Benzoyladenosine Hydrate |
| 153°C | |
| C17H17N5O5 | |
| 1 g | |
| OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12 | |
| 371.35 | |
| 371.35 | |
| Crystalline Powder |
Safety and Handling
TSCA : No