missing translation for 'onlineSavingsMsg'
Learn More

N6-Benzoyladenosine Hydrate 96.0+%, TCI America™

Catalog No. B30931G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30931G 1 g
B30935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30931G Supplier TCI America Supplier No. B30931G

Chemical Identifiers

CAS 4546-55-8
Molecular Formula C17H17N5O5
Molecular Weight (g/mol) 371.35
MDL Number MFCD00056988
InChI Key NZDWTKFDAUOODA-UCQOUBKUNA-N
PubChem CID 11728391
IUPAC Name N-{9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12

Specifications

Melting Point 153°C
Color White
Quantity 1 g
Formula Weight 371.35
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N6-Benzoyladenosine Hydrate