missing translation for 'onlineSavingsMsg'
Learn More

N4-Benzoylcytidine 98.0+%, TCI America™

Catalog No. B30941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30941G 1 g
B30945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30941G Supplier TCI America Supplier No. B30941G

Chemical Identifiers

CAS 13089-48-0
Molecular Formula C16H17N3O6
Molecular Weight (g/mol) 347.33
MDL Number MFCD00010572,MFCD01457402
InChI Key BNXBRFDWSPXODM-UHFFFAOYNA-N
Synonym n6-benzoylcytidine, n-1-2r,4r,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-2-oxopyrimidin-4-yl benzamide
PubChem CID 133611947
IUPAC Name N-{1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide
SMILES OCC1OC(C(O)C1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O

Specifications

Melting Point 240°C
Color White
Quantity 1 g
Formula Weight 347.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N4-Benzoylcytidine