missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N4-Benzoylcytidine, 99%

Catalog No. AAJ6579006
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6579006 5 g
AAJ6579009 10 g
AAJ6579014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6579006 Supplier Thermo Scientific Chemicals Supplier No. J6579006

CAS: 13089-48-0 | C16H17N3O6 | 347.33 g/mol

N-Benzoylcytidine is a useful building block for oligoribonucleotide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoylcytidine is a useful building block for oligoribonucleotide synthesis.

Solubility
Soluble in 1N NaOH at 10mg/ml. Also soluble in methanol or DMSO

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 13089-48-0
Molecular Formula C16H17N3O6
Molecular Weight (g/mol) 347.33
MDL Number MFCD00010572,MFCD01457402
InChI Key BNXBRFDWSPXODM-UHFFFAOYNA-N
Synonym n6-benzoylcytidine, n-1-2r,4r,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-2-oxopyrimidin-4-yl benzamide
PubChem CID 133611947
IUPAC Name N-[1-[(2S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide
SMILES OCC1OC(C(O)C1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O

Specifications

Melting Point 230°C to 234°C (decomposition)
Color White
Quantity 5 g
Solubility Information Soluble in 1N NaOH at 10mg/ml; Also soluble in methanol or DMSO
Optical Rotation +55° (c=0.1 in Methanol)
Formula Weight 347.32
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N4-Benzoylcytidine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only