missing translation for 'onlineSavingsMsg'
Learn More

N2-Isobutyryl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyguanosine 99.0+%, TCI America™

Catalog No. p-7131549
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06971G 1 g
I06975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06971G Supplier TCI America Supplier No. I06971G

Chemical Identifiers

CAS 68892-41-1
Molecular Formula C35H37N5O7
Molecular Weight (g/mol) 639.709
MDL Number MFCD00010059
InChI Key RMQXDNUKLIDXOS-ZGIBFIJWSA-N
Synonym n2-isobutyryl-5'-o-4,4'-dimethoxytrityl-2'-deoxyguanosine, ibu-dmt-dg, dmt-n-ib-dg, 2'-deoxy-5'-o-dmt-n2-isobutyrylguanosine, 5'-o-dimethoxytrityl-n-isobutyryl-deoxyguanosine, guanosine, 5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy-n-2-methyl-1-oxopropyl, n-9-2r,4s,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxyoxolan-2-yl-6-oxo-1h-purin-2-yl-2-methylpropanamide, ksc912c7f, 5'-o-bis 4-methoxyphenyl benzyl-2'-deoxy-n-2-methyl-1-oxopropyl guanosine
PubChem CID 2724504
IUPAC Name N-[9-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide
SMILES CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C3CC(C(O3)COC(C4=CC=CC=C4)(C5=CC=C(C=C5)OC)C6=CC=C(C=C6)OC)O

Specifications

Melting Point 155°C
Color White
Quantity 1 g
Formula Weight 639.71
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N2-Isobutyryl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyguanosine
EINECSNumber (5)-6804
TSCA Yes