Learn More
N1-(4-Amino-2-methylphenyl)acetamide, 90%, Thermo Scientific™
$93.78 - $93.78
Chemical Identifiers
| CAS | 56891-59-9 |
|---|---|
| Molecular Formula | C9H12N2O |
| Molecular Weight (g/mol) | 164.208 |
| MDL Number | MFCD00276633 |
| InChI Key | GWFPMSIIVJMYRZ-UHFFFAOYSA-N |
| Synonym | n-4-amino-2-methylphenyl acetamide, n1-4-amino-2-methylphenyl acetamide, 4'-amino-2'-methylacetanilide, n-4-amino-2-methyl-phenyl-acetamide, 4-acetamido-m-toluidine, 4-acetylamino-m-toluidine, 4-acetamido-3-methylaniline, 4-acetamino-3-methylaniline, acetamide, n-4-amino-2-methylphenyl |
| PubChem CID | 314338 |
| IUPAC Name | N-(4-amino-2-methylphenyl)acetamide |
| SMILES | CC1=C(C=CC(=C1)N)NC(=O)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
BTB06485DA
|
Thermo Scientific Chemicals
BTB06485DA |
1 g | Amber Glass Bottle |
Each for $93.78
|
|
||||
Chemical Identifiers
| 56891-59-9 | |
| 164.208 | |
| GWFPMSIIVJMYRZ-UHFFFAOYSA-N | |
| 314338 | |
| CC1=C(C=CC(=C1)N)NC(=O)C |
| C9H12N2O | |
| MFCD00276633 | |
| n-4-amino-2-methylphenyl acetamide, n1-4-amino-2-methylphenyl acetamide, 4'-amino-2'-methylacetanilide, n-4-amino-2-methyl-phenyl-acetamide, 4-acetamido-m-toluidine, 4-acetylamino-m-toluidine, 4-acetamido-3-methylaniline, 4-acetamino-3-methylaniline, acetamide, n-4-amino-2-methylphenyl | |
| N-(4-amino-2-methylphenyl)acetamide |
Specifications
| 56891-59-9 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD00276633 | |
| n-4-amino-2-methylphenyl acetamide, n1-4-amino-2-methylphenyl acetamide, 4'-amino-2'-methylacetanilide, n-4-amino-2-methyl-phenyl-acetamide, 4-acetamido-m-toluidine, 4-acetylamino-m-toluidine, 4-acetamido-3-methylaniline, 4-acetamino-3-methylaniline, acetamide, n-4-amino-2-methylphenyl | |
| CC1=C(C=CC(=C1)N)NC(=O)C | |
| 164.208 | |
| 164.21 | |
| N1-(4-amino-2-methylphenyl)acetamide |
| 89.5 | |
| 90% | |
| C9H12N2O | |
| 1 g | |
| GWFPMSIIVJMYRZ-UHFFFAOYSA-N | |
| N-(4-amino-2-methylphenyl)acetamide | |
| 314338 | |
| 90% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.