missing translation for 'onlineSavingsMsg'
Learn More

N-(Trifluoromethylthio)phthalimide 98.0+%, TCI America™

Catalog No. p-7137602
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31431G 1 g
T31435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31431G Supplier TCI America Supplier No. T31431G

Chemical Identifiers

CAS 719-98-2
Molecular Formula C9H4F3NO2S
Molecular Weight (g/mol) 247.191
InChI Key CFNSRIIHFLPQCE-UHFFFAOYSA-N
PubChem CID 11334100
IUPAC Name 2-(trifluoromethylsulfanyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)SC(F)(F)F

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 1 g
Formula Weight 247.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(Trifluoromethylthio)phthalimide