accessibility menu, dialog, popup

UserName

CAS RN 719-98-2

ONOSFFF

CAS RN 719-98-2

IUPAC Name: 2-[(trifluoromethyl)sulfanyl]-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-[(trifluoromethyl)sulfanyl]isoindole-1,3-dione
Molecular Weight (g/mol): 247.19
Molecular Formula: C9H4F3NO2S
InChi Key: CFNSRIIHFLPQCE-UHFFFAOYSA-N
SMILES: FC(F)(F)SN1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results