missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)tyramine 97.0+%, TCI America™

Catalog No. B35125G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35125G Supplier TCI America Supplier No. B35125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64318-28-1
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD05864730
InChI Key ILNOTKMMDBWGOK-UHFFFAOYSA-N
Synonym N-Boc-tyramine, N-(tert-Butoxycarbonyl)-4-hydroxyphenethylamine, N-Boc-4-hydroxyphenethylamine
PubChem CID 11831452
IUPAC Name tert-butyl N-[2-(4-hydroxyphenyl)ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCCC1=CC=C(C=C1)O

Specifications

Melting Point 72°C
Quantity 5 g
Formula Weight 237.30
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)tyramine