accessibility menu, dialog, popup

UserName

CAS RN 64318-28-1

H3CH3CCH3OONHOH

CAS RN 64318-28-1

IUPAC Name: tert-butyl N-[2-(4-hydroxyphenyl)ethyl]carbamate
Synonyms: tert-butyl N-[2-(4-hydroxyphenyl)ethyl]carbamate
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: ILNOTKMMDBWGOK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCC1=CC=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results