missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)propargylamine 98.0+%, TCI America™
Chemical Identifiers
| CAS | 92136-39-5 |
|---|---|
| Molecular Formula | C8H13NO2 |
| Molecular Weight (g/mol) | 155.197 |
| MDL Number | MFCD07367245 |
| InChI Key | DSPYCWLYGXGJNJ-UHFFFAOYSA-N |
| Synonym | n-boc-propargylamine, n-tert-butoxycarbonyl propargylamine, n-boc-prop-2-ynylamine, tert-butyl prop-2-yn-1-ylcarbamate, n-boc-propargyl amine, tert-butyl n-prop-2-yn-1-yl carbamate, tert-butyl prop-2-ynylcarbamate, n-boc-2-propyn-1-amine, n-boc propargyl amine, prop-2-ynyl-carbamic acid tert-butyl ester |
| PubChem CID | 11073648 |
| IUPAC Name | tert-butyl N-prop-2-ynylcarbamate |
| SMILES | CC(C)(C)OC(=O)NCC#C |
Chemical Identifiers
| 92136-39-5 | |
| 155.197 | |
| DSPYCWLYGXGJNJ-UHFFFAOYSA-N | |
| 11073648 | |
| CC(C)(C)OC(=O)NCC#C |
| C8H13NO2 | |
| MFCD07367245 | |
| n-boc-propargylamine, n-tert-butoxycarbonyl propargylamine, n-boc-prop-2-ynylamine, tert-butyl prop-2-yn-1-ylcarbamate, n-boc-propargyl amine, tert-butyl n-prop-2-yn-1-yl carbamate, tert-butyl prop-2-ynylcarbamate, n-boc-2-propyn-1-amine, n-boc propargyl amine, prop-2-ynyl-carbamic acid tert-butyl ester | |
| tert-butyl N-prop-2-ynylcarbamate |
Specifications
| 42°C | |
| 170°C | |
| C8H13NO2 | |
| n-boc-propargylamine, n-tert-butoxycarbonyl propargylamine, n-boc-prop-2-ynylamine, tert-butyl prop-2-yn-1-ylcarbamate, n-boc-propargyl amine, tert-butyl n-prop-2-yn-1-yl carbamate, tert-butyl prop-2-ynylcarbamate, n-boc-2-propyn-1-amine, n-boc propargyl amine, prop-2-ynyl-carbamic acid tert-butyl ester | |
| CC(C)(C)OC(=O)NCC#C | |
| 155.197 | |
| 155.20 | |
| Crystalline Powder |
| 92136-39-5 | |
| White-Yellow | |
| MFCD07367245 | |
| DSPYCWLYGXGJNJ-UHFFFAOYSA-N | |
| tert-butyl N-prop-2-ynylcarbamate | |
| 11073648 | |
| ≥98.0% (GC) | |
| N-(tert-Butoxycarbonyl)propargylamine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator