missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)propargylamine 98.0+%, TCI America™

Catalog No. B44725G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44725G 5 g
B44721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44725G Supplier TCI America Supplier No. B44725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 92136-39-5
Molecular Formula C8H13NO2
Molecular Weight (g/mol) 155.197
MDL Number MFCD07367245
InChI Key DSPYCWLYGXGJNJ-UHFFFAOYSA-N
Synonym n-boc-propargylamine, n-tert-butoxycarbonyl propargylamine, n-boc-prop-2-ynylamine, tert-butyl prop-2-yn-1-ylcarbamate, n-boc-propargyl amine, tert-butyl n-prop-2-yn-1-yl carbamate, tert-butyl prop-2-ynylcarbamate, n-boc-2-propyn-1-amine, n-boc propargyl amine, prop-2-ynyl-carbamic acid tert-butyl ester
PubChem CID 11073648
IUPAC Name tert-butyl N-prop-2-ynylcarbamate
SMILES CC(C)(C)OC(=O)NCC#C

Specifications

Melting Point 42°C
Boiling Point 170°C
Color White-Yellow
Quantity 5 g
Formula Weight 155.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)propargylamine
TSCA No
Recommended Storage Refrigerator