accessibility menu, dialog, popup

UserName

CAS RN 92136-39-5

IUPAC Name: tert-butyl N-(prop-2-yn-1-yl)carbamate
Synonyms: tert-butyl N-(prop-2-yn-1-yl)carbamate
Molecular Weight (g/mol): 155.2
Molecular Formula: C8H13NO2
InChi Key: DSPYCWLYGXGJNJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC#C
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)