missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-p-toluenesulfonamide 98.0+%, TCI America™

Catalog No. B164810G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B164810G 10 g
B164825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B164810G Supplier TCI America Supplier No. B164810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18303-04-3
Molecular Formula C12H17NO4S
Molecular Weight (g/mol) 271.331
MDL Number MFCD00134267
InChI Key DUTLOVSBVBGNDM-UHFFFAOYSA-N
Synonym N-Boc-p-toluenesulfonamide
PubChem CID 688170
IUPAC Name tert-butyl N-(4-methylphenyl)sulfonylcarbamate
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(=O)OC(C)(C)C

Specifications

Melting Point 123°C
Color White
Quantity 10 g
Formula Weight 271.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-p-toluenesulfonamide