missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-N-methyl-2-aminoethanol 98.0+%, TCI America™
$41.77 - $155.69
Chemical Identifiers
| CAS | 57561-39-4 |
|---|---|
| Molecular Formula | C8H17NO3 |
| Molecular Weight (g/mol) | 175.228 |
| MDL Number | MFCD03425857 |
| InChI Key | RFDSJHHLGFFVHD-UHFFFAOYSA-N |
| Synonym | n-boc-n-methyl-aminoethanol, tert-butyl 2-hydroxyethyl methyl carbamate, boc,me-glycinol, tert-butyl n-2-hydroxyethyl-n-methylcarbamate, 2-n-methyl-n-boc-amino ethanol, 2-n-boc-n-methylamino ethanol, n-boc-n-methylethanolamine, 2-hydroxyethyl methylcarbamic acid 1,1-dimethylethyl ester, carbamic acid, n-2-hydroxyethyl-n-methyl-, 1,1-dimethylethyl ester |
| PubChem CID | 545700 |
| IUPAC Name | tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate |
| SMILES | CC(C)(C)OC(=O)N(C)CCO |
Chemical Identifiers
| 57561-39-4 | |
| 175.228 | |
| RFDSJHHLGFFVHD-UHFFFAOYSA-N | |
| 545700 | |
| CC(C)(C)OC(=O)N(C)CCO |
| C8H17NO3 | |
| MFCD03425857 | |
| n-boc-n-methyl-aminoethanol, tert-butyl 2-hydroxyethyl methyl carbamate, boc,me-glycinol, tert-butyl n-2-hydroxyethyl-n-methylcarbamate, 2-n-methyl-n-boc-amino ethanol, 2-n-boc-n-methylamino ethanol, n-boc-n-methylethanolamine, 2-hydroxyethyl methylcarbamic acid 1,1-dimethylethyl ester, carbamic acid, n-2-hydroxyethyl-n-methyl-, 1,1-dimethylethyl ester | |
| tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate |
Specifications
| 57561-39-4 | |
| Yellow | |
| MFCD03425857 | |
| RFDSJHHLGFFVHD-UHFFFAOYSA-N | |
| tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate | |
| 545700 | |
| ≥98.0% (GC) | |
| N-(tert-Butoxycarbonyl)-N-methyl-2-aminoethanol |
| 84°C | |
| C8H17NO3 | |
| n-boc-n-methyl-aminoethanol, tert-butyl 2-hydroxyethyl methyl carbamate, boc,me-glycinol, tert-butyl n-2-hydroxyethyl-n-methylcarbamate, 2-n-methyl-n-boc-amino ethanol, 2-n-boc-n-methylamino ethanol, n-boc-n-methylethanolamine, 2-hydroxyethyl methylcarbamic acid 1,1-dimethylethyl ester, carbamic acid, n-2-hydroxyethyl-n-methyl-, 1,1-dimethylethyl ester | |
| CC(C)(C)OC(=O)N(C)CCO | |
| 175.228 | |
| 175.23 | |
| Liquid |
Safety and Handling
TSCA : No