missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-N-methyl-2-aminoethanol 98.0+%, TCI America™

Catalog No. B56855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56855G 5 g
B56851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56855G Supplier TCI America Supplier No. B56855G

Chemical Identifiers

CAS 57561-39-4
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.228
MDL Number MFCD03425857
InChI Key RFDSJHHLGFFVHD-UHFFFAOYSA-N
Synonym n-boc-n-methyl-aminoethanol, tert-butyl 2-hydroxyethyl methyl carbamate, boc,me-glycinol, tert-butyl n-2-hydroxyethyl-n-methylcarbamate, 2-n-methyl-n-boc-amino ethanol, 2-n-boc-n-methylamino ethanol, n-boc-n-methylethanolamine, 2-hydroxyethyl methylcarbamic acid 1,1-dimethylethyl ester, carbamic acid, n-2-hydroxyethyl-n-methyl-, 1,1-dimethylethyl ester
PubChem CID 545700
IUPAC Name tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate
SMILES CC(C)(C)OC(=O)N(C)CCO

Specifications

Boiling Point 84°C
Color Yellow
Quantity 5 g
Formula Weight 175.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-N-methyl-2-aminoethanol