accessibility menu, dialog, popup

UserName

CH3CH3H3COONCH3HO

CAS RN 57561-39-4

IUPAC Name: tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate
Synonyms: tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate
Molecular Weight (g/mol): 175.23
Molecular Formula: C8H17NO3
InChi Key: RFDSJHHLGFFVHD-UHFFFAOYSA-N
SMILES: CN(CCO)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)