missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-D-alanine 98.0+%, TCI America™
$43.67 - $96.84
Chemical Identifiers
| CAS | 7764-95-6 |
|---|---|
| Molecular Formula | C8H15NO4 |
| Molecular Weight (g/mol) | 189.21 |
| MDL Number | MFCD00063123 |
| InChI Key | QVHJQCGUWFKTSE-RXMQYKEDSA-N |
| Synonym | boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid |
| PubChem CID | 637606 |
| IUPAC Name | (2R)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid |
| SMILES | C[C@@H](NC(=O)OC(C)(C)C)C(O)=O |
Chemical Identifiers
| C8H15NO4 | |
| MFCD00063123 | |
| boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid | |
| (2R)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid |
Specifications
| 84°C | |
| White | |
| MFCD00063123 | |
| QVHJQCGUWFKTSE-RXMQYKEDSA-N | |
| (2R)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid | |
| 637606 | |
| ≥98.0% (HPLC,T) | |
| N-(tert-Butoxycarbonyl)-D-alanine |
| 7764-95-6 | |
| C8H15NO4 | |
| boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid | |
| C[C@@H](NC(=O)OC(C)(C)C)C(O)=O | |
| 189.21 | |
| 189.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No