missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-alanine 98.0+%, TCI America™

Supplier:  TCI America B217525G

Quantity 25 g

 View more versions of this product

Catalog No. B217525G

Only null left

Description

Description

Specifications

Chemical Identifiers

7764-95-6
189.21
QVHJQCGUWFKTSE-RXMQYKEDSA-N
637606
C[C@@H](NC(=O)OC(C)(C)C)C(O)=O
C8H15NO4
MFCD00063123
boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid
(2R)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid

Specifications

84°C
7764-95-6
C8H15NO4
boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid
C[C@@H](NC(=O)OC(C)(C)C)C(O)=O
189.21
637606
≥98.0% (HPLC,T)
N-(tert-Butoxycarbonyl)-D-alanine
White
MFCD00063123
QVHJQCGUWFKTSE-RXMQYKEDSA-N
(2R)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
25 g
189.21
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions