accessibility menu, dialog, popup

UserName

CAS RN 7764-95-6

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 189.21
Molecular Formula: C8H15NO4
InChi Key: QVHJQCGUWFKTSE-RXMQYKEDSA-N
SMILES: C[C@@H](NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)