missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-6-aminohexanoic Acid 95.0+%, TCI America™

Supplier:  TCI America B42755G

 View more versions of this product

Catalog No. B42755G

Only null left

Description

Description

Specifications

Specifications

N-(tert-Butoxycarbonyl)-6-aminohexanoic Acid
41°C
166°C
MFCD00037798
boc-epsilon-acp-oh, boc-6-aminocaproic acid, 6-tert-butoxycarbonyl amino hexanoic acid, boc-6-ahx-oh, 6-boc-amino hexanoic acid, boc-, a-acp-oh, hexanoic acid, 6-1,1-dimethylethoxy carbonyl amino, n-boc-6-aminohexanoic acid, 6-boc-amino caproic acid
CC(C)(C)OC(=O)NCCCCCC(=O)O
231.292
231.29
Crystalline Lumps
6404-29-1
White-Yellow
C11H21NO4
5 g
RUFDYIJGNPVTAY-UHFFFAOYSA-N
6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
637602
≥95.0% (GC,T)

Chemical Identifiers

6404-29-1
231.292
RUFDYIJGNPVTAY-UHFFFAOYSA-N
637602
CC(C)(C)OC(=O)NCCCCCC(=O)O
C11H21NO4
MFCD00037798
boc-epsilon-acp-oh, boc-6-aminocaproic acid, 6-tert-butoxycarbonyl amino hexanoic acid, boc-6-ahx-oh, 6-boc-amino hexanoic acid, boc-, a-acp-oh, hexanoic acid, 6-1,1-dimethylethoxy carbonyl amino, n-boc-6-aminohexanoic acid, 6-boc-amino caproic acid
6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions